Order Entry
Netherlands
ContactUsLinkComponent
NucleoBond PC 500 EF (10) 10 preps for the isolation of endotoxin-free plasmid DNA - NucleoBond AX 500 Columns, NucleoBond Folded Fi lters, buffers, RNase A no dangerous goods according transport regulations 1 * 10 items
Catalog # MANA740550
Supplier:  MACHEREY-NAGEL
undefined
NucleoBond PC 500 EF (10) 10 preps for the isolation of endotoxin-free plasmid DNA - NucleoBond AX 500 Columns, NucleoBond Folded Fi lters, buffers, RNase A no dangerous goods according transport regulations 1 * 10 items
Catalog # MANA740550
Supplier:  MACHEREY-NAGEL
Frequently Bought Together