Order Entry
Netherlands
ContactUsLinkComponent
NucleoSpin Finisher Midi (10) 10 preps for purification of plasmid DNA eluates from NucleoBond PC 100 and Xtra Midi(EF) preparations , processing by centrifugation - 10 NucleoSpin Finisher Columns, buffers, Collection Tubes 1 * 10 items
Catalog # MANA740439.10
Supplier:  MACHEREY-NAGEL
undefined
NucleoSpin Finisher Midi (10) 10 preps for purification of plasmid DNA eluates from NucleoBond PC 100 and Xtra Midi(EF) preparations , processing by centrifugation - 10 NucleoSpin Finisher Columns, buffers, Collection Tubes 1 * 10 items
Catalog # MANA740439.10
Supplier:  MACHEREY-NAGEL
Frequently Bought Together