Order Entry
Netherlands
ContactUsLinkComponent
NucleoSnap DNA Plasma(50) 50 preps for the isolation of circulating DNA from plasma - NucleoSnap DNA Plasma Columns, Collection Tube s, buffers, Proteinase K __UN 1993 Flammable liquid, n.o.s. (ethanol, 2-propanol mixture) 3 III 0.12 1 * 50 items
Catalog # MANA740300.50
Supplier:  MACHEREY-NAGEL
undefined
NucleoSnap DNA Plasma(50) 50 preps for the isolation of circulating DNA from plasma - NucleoSnap DNA Plasma Columns, Collection Tube s, buffers, Proteinase K __UN 1993 Flammable liquid, n.o.s. (ethanol, 2-propanol mixture) 3 III 0.12 1 * 50 items
Catalog # MANA740300.50
Supplier:  MACHEREY-NAGEL
Frequently Bought Together