NucleoSpin 8 RNA Blood (12x8) 12x 8 preps for the isolation of RNA from blood - NucleoSpin RNA Blood Binding Strips, MN Wash Plates, Racks of Tube Strips, Square-well Blocks, Elution Plates U-bottom, Self-adhering Foils, buffers, RNase-free rDNase, 1 * 96 items
MANA740220
Frequently Bought Together