Precision thermometer like ISO-R 1770 L/75, BS 1704, ANSI CP 45, capillary yellow enamelled round, red filling, range: - 1+101°C in 0.1°C Immersion 75 mm, total length: 610 mm, suitable for calibration - each packed in a transparent plastic case 1 * 1 items
Catalog # SCNL1232612S
Supplier: Ludwig Schneider GmbH & CO. KG
Frequently Bought Together