Adapter polyamide for clamping lid for dry sieving with test sieves 200 mm/ 8inches dia. with the Heavy Duty Sieve Shaker ANALYSETTE 18 1 * 1 items
FRIT18.3051.00
: FRITSCH
Adapter polyamide for clamping lid for dry sieving with test sieves 200 mm/ 8inches dia. with the Heavy Duty Sieve Shaker ANALYSETTE 18 1 * 1 items
FRIT18.3051.00
: FRITSCH
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection