Order Entry
Netherlands
ContactUsLinkComponent
Sieve insert 20 mm square perforation made of stainnless steel for Cutting Mill PULVERISETTE 15 null
Catalog # FRIT45.2300.10
Supplier:  FRITSCH
Sieve insert 20 mm square perforation made of stainnless steel for Cutting Mill PULVERISETTE 15 null
Catalog # FRIT45.2300.10
Supplier:  FRITSCH
Frequently Bought Together