Order Entry
Netherlands
ContactUsLinkComponent
Reprosil Pur 120 ODS-3, 3.0 µm, 4 x 100 mm, 17% Carbon,HPLC Column endcapped 1 * 1 items
Catalog # MAISR13.93.S1004
Supplier:  DR MAISCH
Reprosil Pur 120 ODS-3, 3.0 µm, 4 x 100 mm, 17% Carbon,HPLC Column endcapped 1 * 1 items
Catalog # MAISR13.93.S1004
Supplier:  DR MAISCH
Frequently Bought Together