Order Entry
Netherlands
ContactUsLinkComponent
Bimetal dial therm., axial, 0+120:2°C, built-in length 63mm, diameter of case 80mm, thread R 1/2 1 * 1 items
Catalog # AMART110354
Supplier:  Amarell
Bimetal dial therm., axial, 0+120:2°C, built-in length 63mm, diameter of case 80mm, thread R 1/2 1 * 1 items
Catalog # AMART110354
Supplier:  Amarell
Frequently Bought Together