Order Entry
Netherlands
ContactUsLinkComponent
Precision thermometer, similar DIN, 0+100:0,1C, enclosed scale, cap. lens front, red special liquid, Richter-top finish with button , 550x9-9,5mm, with Works Certificate with 5 test points 1 * 1 items
Catalog # AMARL26004-FL/WC
Supplier:  Amarell
Precision thermometer, similar DIN, 0+100:0,1C, enclosed scale, cap. lens front, red special liquid, Richter-top finish with button , 550x9-9,5mm, with Works Certificate with 5 test points 1 * 1 items
Catalog # AMARL26004-FL/WC
Supplier:  Amarell
Frequently Bought Together