Alcoholometer, EUROPA, class I, 80-90 :0,1%vol., without therm., ref. temp. 20C, 400mm long, suitable for government verification 1 * 1 items
AMARH848036
: Amarell
Alcoholometer, EUROPA, class I, 80-90 :0,1%vol., without therm., ref. temp. 20C, 400mm long, suitable for government verification 1 * 1 items
AMARH848036
: Amarell
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection