Order Entry
Netherlands
ContactUsLinkComponent
ASTM-Thermom., S56C, white backed, 19+35:0,02C, capillary tube specially coated inside, with non-wetting blue special liquid, tota l immersion, max.622x7-8mm, durable pigment, null
Catalog # AMARA300750-CB
Supplier:  Amarell
ASTM-Thermom., S56C, white backed, 19+35:0,02C, capillary tube specially coated inside, with non-wetting blue special liquid, tota l immersion, max.622x7-8mm, durable pigment, null
Catalog # AMARA300750-CB
Supplier:  Amarell
Frequently Bought Together