Thermometer similar to BS 1365, SB95C/total, 85+95:0,1°C, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402560-CB
Supplier: Amarell
Frequently Bought Together