Order Entry
Netherlands
ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 7d, solid stem, white backed, 65+90:0,1°C, length 400mm, imm. 100mm, capillary tube specially c oated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400040-CB
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 7d, solid stem, white backed, 65+90:0,1°C, length 400mm, imm. 100mm, capillary tube specially c oated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400040-CB
Supplier:  Amarell
Frequently Bought Together