Order Entry
Netherlands
ContactUsLinkComponent
UMKEHRFRITTE1000ML M. 2XGLASHAHN NS14/2, 5 FRIT D. 80 P4 NS29 OLIVE 8MM L. 350MM FOR FILTRATION OXIDATION AND HYDROLYSIS-MORE SENSITIVE SUBSTANCES BORO 3.3 1 * 1 items
Catalog # NEUB4324-1000-294
UMKEHRFRITTE1000ML M. 2XGLASHAHN NS14/2, 5 FRIT D. 80 P4 NS29 OLIVE 8MM L. 350MM FOR FILTRATION OXIDATION AND HYDROLYSIS-MORE SENSITIVE SUBSTANCES BORO 3.3 1 * 1 items
Catalog # NEUB4324-1000-294
Frequently Bought Together