Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Morpholine 99%

Morpholine 99%

Bottle, 100 g
Catalog # APOSOR13977-100G
Corrosive
Flammable
Irritant
Supplier:   Apollo Scientific
Morpholine 99%
Catalog # APOSOR13977-100G
Supplier:  Apollo Scientific
CAS Number:  110-91-8
Morpholine 99%

Bottle, 100 g

CAS Number:  110-91-8
Price
29,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:110-91-8
  • Formula:C₄H₉NO
  • MDL Number:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Boiling Pt:129 °C
  • Melting Pt:–7…–5 °C
  • Melting Pt:–7 to –5 °C
  • Density:0.996 g/cm³ (25 °C)
  • Flash Pt:31 °C
  • Storage Temperature:Ambient
  • EINECS:203-815-1
  • UN:2054
  • ADR:8+3,I
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.