Order Entry
Netherlands
Orders LinkContactUsLinkComponent
4-Hydroxy-7-methylthieno[3,2-d]pyrimidine

4-Hydroxy-7-methylthieno[3,2-d]pyrimidine

Bottle, 5 g
Catalogus # APOSOR110039-5G
Irriterend
Leverancier:   Apollo Scientific
4-Hydroxy-7-methylthieno[3,2-d]pyrimidine
Catalogus # APOSOR110039-5G
Leverancier:  Apollo Scientific
CAS-nummer:  175137-13-0
4-Hydroxy-7-methylthieno[3,2-d]pyrimidine

Bottle, 5 g

CAS-nummer:  175137-13-0
Price
109,00 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents
Synoniemen:  7-Methylthieno[3,2-d]pyrimidin-4-ol
  • CAS-nummer:175137-13-0
  • Formule:C₇H₆N₂OS
  • MDL-nummer:MFCD13181517
  • Molecular Weight:166.2 g/mol
  • Smelten Pt:240…242 °C
  • Temperatuur bij opslag:Ambient
Specificaties

Specificaties

  • Pack type:
    Bottle
  • Pk:
    5 g

Specificaties

Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.