Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Anderson's Basal salt mixture

Anderson's Basal salt mixture

Anderson's basal salt mixture 50 L, 50 L
Catalog # APOSPMA267C-50L
Supplier:   Apollo Scientific
Anderson's Basal salt mixture
Catalog # APOSPMA267C-50L
Supplier:  Apollo Scientific
CAS Number:  
Anderson's Basal salt mixture

Anderson's basal salt mixture 50 L, 50 L

CAS Number:  
Price
31,90 €
Log in for availability
1 * 50 L
Product Details & Documents

Contains macro/micronutrients as described byAnderson, 1978/80 Plant tissue culture testedFull specification given in appendix.

  • MFCD00243139
Specifications

Specifications

  • Pk:
    50 L
  • Description:
    Anderson's basal salt mixture 50 L

Specifications