Anderson's Basal salt mixture
Anderson's basal salt mixture 10×1 L, 10
Catalog # APOSPMA267A-10X1L
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Contains macro/micronutrients as described byAnderson, 1978/80 Plant tissue culture testedFull specification given in appendix.
- MFCD00243139
Specifications
Specifications
- Pk:10
- Description:Anderson's basal salt mixture 10×1 L