Order Entry
Netherlands
Orders LinkContactUsLinkComponent
2-Furaldehyde

2-Furaldehyde

Bottle, 250 g
Catalog # APOSOR23023-250G
Flammable
Toxic
Health
Supplier:   Apollo Scientific
2-Furaldehyde
Catalog # APOSOR23023-250G
Supplier:  Apollo Scientific
CAS Number:  98-01-1
2-Furaldehyde

Bottle, 250 g

CAS Number:  98-01-1
Price
50,70 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  2-Furfurol, Furfural
  • CAS Number:98-01-1
  • Formula:C₅H₄O₂
  • MDL Number:MFCD00003229
  • Molecular Weight:96.09 g/mol
  • Boiling Pt:159…162 °C
  • Melting Pt:–38 °C
  • Density:1.159 g/cm³ (25 °C)
  • Flash Pt:60 °C
  • Storage Temperature:Ambient
  • EINECS:202-627-7
  • UN:1199
  • ADR:6.1+3,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications