2-Furaldehyde
Bottle, 1 kg
APOSOR23023-1KG
: 2-Furfurol, Furfural
- :98-01-1
- :C₅H₄O₂
- :MFCD00003229
- :96.09 g/mol
- :159…162 °C
- :–38 °C
- :1.159 g/cm³ (25 °C)
- :60 °C
- :Ambient
- :202-627-7
- :1199
- :6.1+3,II
- Pack type:Bottle
- Pk:1 kg