3,5-Dimethylanisole
Bottle, 5 g
Catalog # APOSOR9605-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Methoxy-3,5-dimethylbenzene
- CAS Number:874-63-5
- Formula:C₉H₁₂O
- MDL Number:MFCD00008398
- Molecular Weight:136.19 g/mol
- Boiling Pt:192…193 °C
- Density:0.963 g/cm³ (25 °C)
- Flash Pt:65 °C
- Storage Temperature:Ambient
- EINECS:212-865-3
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g



