Bromo-PEG3-t-butyl ester
Bottle, 250 mg
Catalog # APOSBIPG1349-250MG
Supplier: Apollo Scientific
Bromo-PEG3-t-butyl ester
Catalog # APOSBIPG1349-250MG
Supplier: Apollo Scientific
CAS Number: 782475-37-0
Jump to:
Product Details & Documents
Synonyms: Br-PEG3-C2-Boc
- CAS Number:782475-37-0
- Formula:C₁₃H₂₅BrO₅
- MDL Number:MFCD22574821
- Molecular Weight:341.239 g/mol
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



