Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Doxycycline hydrochloride

Doxycycline hydrochloride

Bottle, 10 g
Catalog # APOSBID0121-10G
Irritant
Supplier:   Apollo Scientific
Doxycycline hydrochloride
Catalog # APOSBID0121-10G
Supplier:  Apollo Scientific
CAS Number:  10592-13-9
Doxycycline hydrochloride

Bottle, 10 g

CAS Number:  10592-13-9
Price
37,70 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  (4S,4aR,5S,5aR,6R,12aS)-4-(Dimethylamino)-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
  • CAS Number:10592-13-9
  • Formula:C₂₂H₂₄N₂O₁₀·HCl
  • MDL Number:MFCD03427564
  • Molecular Weight:480.9 g/mol
  • Melting Pt:210 °C
  • Storage Temperature:Refrigerator
  • EINECS:234-198-7

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.