Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

2,6-Difluorophenylhydrazine

Bottle
Catalogus # APOSPC1502-5G
Leverancier:   Apollo Scientific
2,6-Difluorophenylhydrazine
Catalogus # APOSPC1502-5G
Leverancier:  Apollo Scientific
CAS-nummer:  119452-66-3
2,6-Difluorophenylhydrazine

Bottle

CAS-nummer:  119452-66-3
Price
222,00 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents
Synoniemen:  (2,6-Difluorophenyl)hydrazine
  • CAS-nummer:119452-66-3
  • Formule:C₆H₆F₂N₂
  • MDL-nummer:MFCD06658266
  • Molecular Weight:144.122 g/mol
  • Temperatuur bij opslag:Ambient
Specificaties

Specificaties

  • Pack type:
    Bottle

Specificaties

Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.