Order Entry
Netherlands
ContactUsLinkComponent
Fixed high intensity multiplexing LEDs with peak band passes of Red (635 nm), Green (520 nm), Blue (465 nm), Infra-Red (740 nm) 1 * 1 items
Catalog # SYNOGM-HILED-RGBIR
Fixed high intensity multiplexing LEDs with peak band passes of Red (635 nm), Green (520 nm), Blue (465 nm), Infra-Red (740 nm) 1 * 1 items
Catalog # SYNOGM-HILED-RGBIR
Frequently Bought Together