Order Entry
Netherlands
ContactUsLinkComponent
CRAMPS PTFE spacer for BL4 MAS rotor only, outer diameter 4.0mm, PTFE, temperature range +10 to +50C 1 * 1 items
Catalog # BRUKHZ1872
Supplier:  BRUKER DALTRONIK
CRAMPS PTFE spacer for BL4 MAS rotor only, outer diameter 4.0mm, PTFE, temperature range +10 to +50C 1 * 1 items
Catalog # BRUKHZ1872
Supplier:  BRUKER DALTRONIK
Frequently Bought Together