SteriWare® Liquid Funnels
Funnel, natural, 11 mm, 100 mm
Catalog # 300-0592
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Single use, disposable liquid funnels ideal for laboratories and manufacturing areas. Designed to use straight from bags.
- Made of HDPE
- Moulded and packed in a class 100,000 cleanroom
- With small hygenic side handle
- BSE/TSE free
- Available sterile (gamma irradiated)
: Material conforms to FDA, EU 10/2011 and EC 1935/2004. Full batch traceability.
: Individually bagged.
Specifications
- Description:Funnel, natural
- Seq:05
- Int.Ø top:100 mm
- Stem O-Ø:11 mm



