Order Entry
Netherlands
ContactUsLinkComponent
Easy Purchase Pack 2ml clear RSA-Pro glass vials (12x32mm) w/ write on patch and blue Soft-Guard screw caps w/ bonded silicone/PTFE septa in clear kit pack. AQ brand. 100/pack. 1 * 100 items
Catalog # MICS9509S-1WDC-EMR
Supplier:  MICROSOLV
undefined
Easy Purchase Pack 2ml clear RSA-Pro glass vials (12x32mm) w/ write on patch and blue Soft-Guard screw caps w/ bonded silicone/PTFE septa in clear kit pack. AQ brand. 100/pack. 1 * 100 items
Catalog # MICS9509S-1WDC-EMR
Supplier:  MICROSOLV
Frequently Bought Together