Lead Standard Solution, CRM traceable to SRM from NIST 0.100 mg/l Pb in HNO3 1 * 100 mL
Catalog # 1.33004.0100
Supplier: Merck
Lead Standard Solution, CRM traceable to SRM from NIST 0.100 mg/l Pb in HNO3 1 * 100 mL
Catalog # 1.33004.0100
Supplier: Merck
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
KCL ELECTROLYTE SOLUTION CPY4 3.0 MOL 1 * 250 mL
Supplier: Endress+Hauser
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
pH buffers, ready to use
Supplier: WTW