Order Entry
Netherlands
ContactUsLinkComponent
In Pro InPro 6820, diam. 25 mm T-82 Connector Immersion length: 160 mm Accuracy,,+/-1 % +6 ppb Sterilizable,, Autoclavable,, Response time at 25 C (air --> N2),,90 sec. Mechanical pressure resistance,,max. 12 bar (174 psi) absolute Detection Range,, 1 * 1 items
Catalog # METN52201021
Supplier:  Mettler - Toledo
In Pro InPro 6820, diam. 25 mm T-82 Connector Immersion length: 160 mm Accuracy,,+/-1 % +6 ppb Sterilizable,, Autoclavable,, Response time at 25 C (air --> N2),,90 sec. Mechanical pressure resistance,,max. 12 bar (174 psi) absolute Detection Range,, 1 * 1 items
Catalog # METN52201021
Supplier:  Mettler - Toledo
Frequently Bought Together