Iron Standard Solution, CRM traceable to SRM from NIST 0.300 mg/l Fe(III) in HNO3 1 * 100 mL
Catalog # 1.33019.0100
Supplier: Merck
Iron Standard Solution, CRM traceable to SRM from NIST 0.300 mg/l Fe(III) in HNO3 1 * 100 mL
Catalog # 1.33019.0100
Supplier: Merck
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Water, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals