Iron Standard Solution, CRM traceable to SRM from NIST 0.1000 mg/l Fe(III) in HNO3 1 * 100 mL
Catalog # 1.33018.0100
Supplier: Merck
Iron Standard Solution, CRM traceable to SRM from NIST 0.1000 mg/l Fe(III) in HNO3 1 * 100 mL
Catalog # 1.33018.0100
Supplier: Merck
Frequently Bought Together
Sterilin™ Quickstart Universal Containers, PS, 30 ml
Supplier: STERLIN
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Pipette tips, PIPETMAN® Diamond®
Supplier: GILSON
VWR® Bottles, Water Sampling
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Hardness quality control standard
Supplier: Hach