Bestelling invoeren
Netherlands
ContactUsLinkComponent
Iron Standard Solution, CRM traceable to SRM from NIST 0.0500 mg/l Fe(III) in HNO3 1 * 100 mL
Catalog # 1.33014.0100
Supplier:  Merck
Iron Standard Solution, CRM traceable to SRM from NIST 0.0500 mg/l Fe(III) in HNO3 1 * 100 mL
Catalog # 1.33014.0100
Supplier:  Merck