Chromium Standard Solution, CRM traceable to SRM from NIST 0.050 mg/l Cr(VI) in H2O 1 * 100 mL
Catalog # 1.33012.0100
Supplier: Merck
Chromium Standard Solution, CRM traceable to SRM from NIST 0.050 mg/l Cr(VI) in H2O 1 * 100 mL
Catalog # 1.33012.0100
Supplier: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Nitrogen (total) CRM, Standard Solutions, Supelco®
Supplier: Merck
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection