Chloride Standard Solution, CRM traceable to SRM from NIST 1.00 mg/l Cl¯ in H2O 1 * 100 mL
Catalog # 1.33010.0100
Supplier: Merck
Chloride Standard Solution, CRM traceable to SRM from NIST 1.00 mg/l Cl¯ in H2O 1 * 100 mL
Catalog # 1.33010.0100
Supplier: Merck
Frequently Bought Together
Potassium pyroantimonate 0.05 mol/l in potassium hydroxide solution Reag. Ph. Eur. 1071301
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals