Chromium Standard Solution, CRM traceable to SRM from NIST, 1.00 mg/l Cr(VI) in H2O 1 * 100 mL
Catalog # 1.33013.0100
Supplier: Merck
Chromium Standard Solution, CRM traceable to SRM from NIST, 1.00 mg/l Cr(VI) in H2O 1 * 100 mL
Catalog # 1.33013.0100
Supplier: Merck
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Visual colour kits, ammonium, MQuant®, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Glass microfibre filters without binder, grade GF/C RTU, Whatman™
Supplier: Whatman products (Cytiva)
SGE BPX1 GC Capillary Columns
Supplier: Trajan Scientific and Medical
Chromium standard solutions
Supplier: Hach
Portable balances, Compass™ CR series
Supplier: OHAUS