Order Entry
Netherlands
Orders LinkContactUsLinkComponent
STEMdiff™ Neural Induction Medium
STEMdiff™ Neural induction medium, 250 mL
  STMC05835
Marketsource
STEMdiff™ Neural Induction Medium
  STMC05835
Marketsource
 :  
STEMdiff™ Neural Induction Medium
STEMdiff™ Neural Induction Medium
STEMdiff™ Neural induction medium, 250 mL
 :  
328,00 €
1 * 250 mL
Defined, serum-free medium for neural induction of human ES and iPS cells.
STEMdiff™ Neural Induction Medium is a defined, serum-free medium for the neural induction of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. This medium enables highly efficient generation of neural progenitor cell using either embryoid body- or monolayer culture-based protocols.Learn how to generate neural progenitor cells from human pluripotent stem cells (hPSCs) in our On-Demand Neural Induction Course, and browse our Tech Tips on the Neural Induction of hPSCs using the Embryoid body method or monolayer method.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
  • Pk:
    250 mL
  • Description:
    STEMdiff™ Neural induction medium