Order Entry
Netherlands
ContactUsLinkComponent
CONTAINS ACCESSORIES, CONSUMABLES AND APPLICATION SOFTWARE TO PERFORM OIL IN WATER MEASUREMENTS BY EXTRACTION INTO CYCLOHEXANE THEN TRANSMISSION FT-IR.COMPRISES: 2 PORT LUER LOCK %T CELL - CAF2 0.5MM 2 X 2ML LUER LOCK SYRINGE SPARE 0.5MM PTFE SPACER 1 * 1 items
Catalog # PERCL1608007
Supplier:  PerkinElmer
CONTAINS ACCESSORIES, CONSUMABLES AND APPLICATION SOFTWARE TO PERFORM OIL IN WATER MEASUREMENTS BY EXTRACTION INTO CYCLOHEXANE THEN TRANSMISSION FT-IR.COMPRISES: 2 PORT LUER LOCK %T CELL - CAF2 0.5MM 2 X 2ML LUER LOCK SYRINGE SPARE 0.5MM PTFE SPACER 1 * 1 items
Catalog # PERCL1608007
Supplier:  PerkinElmer
Frequently Bought Together