A BLACK, ROUND, METAL SAMPLE CUP WITH QUARTZ GLASS INSTALLED IN THE BASE. THE CUP COMES WITH A LOCKING METAL BACK AND A FOAM INSERT. THE CUP CAN BE USED TO ANALYZE DRY, GROUND FOOD OR AGRICULTURE PRODUCTS. ONCE THE CUP IS FILLED, PLEASE USE DISPOSAB 1 * 1 items
Catalog # PERCL1281933
Supplier: PerkinElmer
Frequently Bought Together