Order Entry
Netherlands
ContactUsLinkComponent
CALIBRATED 99% REFLECTIVE SINTERED PTFE POLYMER DISC 8 MM THICK X 58 MM DIAMETER WITHIN A SEALABLE CONTAINER SUITABLE FOR THE NIRA I I 60 MM SPINNER. 1 * 1 items
Catalog # PERCL1281922
Supplier:  PerkinElmer
CALIBRATED 99% REFLECTIVE SINTERED PTFE POLYMER DISC 8 MM THICK X 58 MM DIAMETER WITHIN A SEALABLE CONTAINER SUITABLE FOR THE NIRA I I 60 MM SPINNER. 1 * 1 items
Catalog # PERCL1281922
Supplier:  PerkinElmer
Frequently Bought Together