MAG BIND PARTICLES HDQ 1 * 50 mL
MBPHDQ-50
Frequently Bought Together
VWR®, Microscope Slides
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Disinfectant solutions, Bacillol AF
: BODE CHEMIE
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Formalin 10%, Q Path®, buffered
: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Cooling Bricks
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF