Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Disodium hydrogen phosphate

Disodium hydrogen phosphate

250 g
Catalogus # 5.43838.0250
Leverancier:   Merck
Disodium hydrogen phosphate
Catalogus # 5.43838.0250
Leverancier:  Merck
CAS-nummer:  7558-79-4
Disodium hydrogen phosphate

250 g

CAS-nummer:  7558-79-4
Price
834,88 €
Log in voor beschikbaarheid
1 * 250 g
Product Details & Documents
Synoniemen:  sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
  • CAS-nummer:7558-79-4
  • Formule:Na₂HPO₄
  • MDL-nummer:MFCD00003496
  • Molecular Weight:141.96 g/mol
  •  :M-Clarity™ quality level = MQ100
Specificaties

Specificaties

  • Pk:
    250 g

Specificaties

Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.