REF. ELECTRODE FILLING SOLUTION(4X5ML) 1 * 20 mL
MSPP2563698
Frequently Bought Together
Nitenpyram Analytical standard, reference material, Fluka™
: Honeywell Chemicals
CEFAZOLIN SODIUM SALT 1 * 75 mg
: Merck
TEPRALOXYDIM CERTIFIED 1 * 0,1 g
: EHRENSTORFER
PIRACETAM 1 * 500 mg
: Merck
RAPIZYME TRYPSIN, MS GRADE, 4/P
: WATERS
AMMONIUM CAL SOLUTION 10MG/L 1 * 500 mL
: YSI ENVIRONMENTAL
FONOFOS CERTIFIED 1 * 0,1 g
: EHRENSTORFER
VWR®, Flow Cup viscosity standards
: VWR Chemicals