Order Entry
Netherlands
ContactUsLinkComponent
sodium phosphate dibasic solution [na2hpo4] is used to prepare phosphate based biological buffers which are used in molecular biolog y, biochemistry and chromatography 1 * 100 mL
Catalog # GENO786-1203
Supplier:  G-Biosciences
sodium phosphate dibasic solution [na2hpo4] is used to prepare phosphate based biological buffers which are used in molecular biolog y, biochemistry and chromatography 1 * 100 mL
Catalog # GENO786-1203
Supplier:  G-Biosciences
Frequently Bought Together