Order Entry
Netherlands
ContactUsLinkComponent
sodium phosphate monobasic solution [nah2po4] is used to prepare phosphate based biological buffers which are used in molecular biol ogy, biochemistry and chromatography 1 * 500 mL
Catalog # GENO786-1202
Supplier:  G-Biosciences
sodium phosphate monobasic solution [nah2po4] is used to prepare phosphate based biological buffers which are used in molecular biol ogy, biochemistry and chromatography 1 * 500 mL
Catalog # GENO786-1202
Supplier:  G-Biosciences
Frequently Bought Together