Order Entry
Netherlands
ContactUsLinkComponent
jaw borate buffer packs are dry blend powder pouches used to make borate buffer which is used in protein modification procedures th at require amine free and alkaline ph conditions 1 * 1 KIT
Catalog # GENO786-1207
Supplier:  G-Biosciences
jaw borate buffer packs are dry blend powder pouches used to make borate buffer which is used in protein modification procedures th at require amine free and alkaline ph conditions 1 * 1 KIT
Catalog # GENO786-1207
Supplier:  G-Biosciences
Frequently Bought Together