SODIUM BISULFITE SOLUTION 30% RECTAPUR 1 * 500 mL
Catalog # PRLS89772.260
Supplier: PROLABO CMP
Frequently Bought Together
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
METHYLENE BLUE RAL 555 AQUEOUS SOL. 1L 1 * 1 L
Supplier: RAL
BOND EPITOPE RETRIEVAL SOLUTION 2 1 * 1 L
Supplier: LEICA MICROSYSTEMS
[EN]TECHNOVIT 9100 REGULATOR 1 X 5 ML 1 * 5 mL
Supplier: HERAEUS KULZER
NANOCOLOR® Photometric Tube Tests, Chromium
Supplier: MACHEREY-NAGEL
Reagents for ammonia determination using Lovibond® MD photometers
Supplier: Lovibond
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS
Nitrite VHR L, 500 ml
Supplier: Lovibond
VWR® Q Path® O.C.T. Compound Mounting Medium for Cryotomy
Supplier: VWR Chemicals