Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Bottle-top dispenser VITLAB® Simplex², Volume: 1.0 - 10.0 ml, DE-M marked 1 * 1 items
Bottle-top dispenser VITLAB® Simplex², Volume: 1.0 - 10.0 ml, DE-M marked 1 * 1 items
Catalog # VITL1621505
Supplier:   VITLAB
Bottle-top dispenser VITLAB® Simplex², Volume: 1.0 - 10.0 ml, DE-M marked 1 * 1 items
Catalog # VITL1621505
Supplier:  VITLAB
Bottle-top dispenser VITLAB® Simplex², Volume: 1.0 - 10.0 ml, DE-M marked 1 * 1 items
Price
436,00 €
Log in for availability
1 * 1 ST
Product Details & Documents