Order Entry
Netherlands
ContactUsLinkComponent
AgriseraECL SuperBright (100 ml), 2 x 50 ml, two component ready to use solutions, enough for 50 midi blots (6.8 x 8.1 cm) 1 * 100 mL
Catalog # AGRIAS16ECL-S-100
Supplier:  Agrisera
AgriseraECL SuperBright (100 ml), 2 x 50 ml, two component ready to use solutions, enough for 50 midi blots (6.8 x 8.1 cm) 1 * 100 mL
Catalog # AGRIAS16ECL-S-100
Supplier:  Agrisera